Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- 5033424D13Rik
- Entrez Gene ID
-
75975
- Gene Name
- RIKEN cDNA 5033424D13 gene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017190.1 |
- |
1 - 1 |
546 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY716920.1 |
- |
1 |
542 |
|
Chr. 12 - 108878772 - 108879418 [+] on Build GRCm38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tryptophanyl-tRNA synthetase |
Wars |
TrpRS, WRS |
|
Anti-sense |
Chr. - 108860030 - 108894174 on Build GRCm38 |
Downstream |
WD repeat domain 25 |
Wdr25 |
B930090D16Rik |
14.8kb
|
Sense |
Chr. - 108894228 - 109028452 on Build GRCm38 |
View Full Product Details