Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plxdc2
Entrez Gene ID
67448
Gene Name
plexin domain containing 2
Gene Aliases
1200007L24Rik, 5430431D22Rik, AI646728, AU022916, AV231634, Tem7r
Chromosome Location
Chr.2: 16356210 - 16755839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026162.6 NP_080438.2 14 - 14 6010 91
GenBank mRNA AK017369.1 - 1 - 1 1154 91

View Full Product Details