Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB750900.1 |
- |
1 |
|
|
Chr. 12 - 73040188 - 73040609 [+] on Build GRCm38 |
103 |
GenBankmRNA |
BQ920331.1 |
- |
1 |
373 |
|
Chr. 12 - 73039952 - 73040849 [-] on Build GRCm38 |
103 |
GenBankmRNA |
BQ887071.1 |
- |
2 |
401 |
|
Chr. 12 - 73040015 - 73042815 [-] on Build GRCm38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sine oculis-related homeobox 6 |
Six6 |
Optx2, Six9 |
-95.1kb
|
Anti-sense |
Chr. - 72939945 - 72944899 on Build GRCm38 |
Downstream |
sine oculis-related homeobox 1 |
Six1 |
BB138287 |
1.3kb
|
Sense |
Chr. - 73041827 - 73046712 on Build GRCm38 |
View Full Product Details