Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddi2
Entrez Gene ID
68817
Gene Name
DNA-damage inducible protein 2
Gene Aliases
1110056G13Rik, 1700027M01Rik, 9130022E05Rik, AI604911, AU040698
Chromosome Location
Chr.4: 141683563 - 141723419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK165215.1 - - 2554 103


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA564951.1 - 1 131 Chr. 4 - 141679043 - 141679551 [-] on Build GRCm38 108
GenBankmRNA BF460839.1 - 1 144 Chr. 4 - 141679279 - 141679567 [-] on Build GRCm38 103
GenBankmRNA BE957335.1 - 1 143 Chr. 4 - 141678990 - 141679567 [-] on Build GRCm38 103
GenBankmRNA CO424225.1 - 1 180 Chr. 4 - 141678921 - 141679567 [-] on Build GRCm38 103
GenBankmRNA CB205302.1 - 378 Chr. - - [-] on Build 103
GenBankmRNA CF730295.1 - 1 183 Chr. 4 - 141678942 - 141679567 [-] on Build GRCm38 103
GenBankmRNA CB522283.1 - 1 182 Chr. 4 - 141678946 - 141679573 [-] on Build GRCm38 103
GenBankmRNA CA564361.1 - 1 131 Chr. 4 - 141679115 - 141679551 [-] on Build GRCm38 108
GenBankmRNA BE688076.1 - 1 126 Chr. 4 - 141679082 - 141679551 [-] on Build GRCm38 103
GenBankmRNA CJ161633.1 - 1 175 Chr. 4 - 141679211 - 141679581 [-] on Build GRCm38 103


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream pleckstrin homology domain containing, family M (with RUN domain) member 2 Plekhm2 2310034J19Rik, AI854247, mKIAA0842 -14.9kb Sense Chr. - 141625732 - 141664151 on Build GRCm38

View Full Product Details