Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Mpzl1
- Entrez Gene ID
-
68481
- Gene Name
- myelin protein zero-like 1
- Gene Aliases
-
1110007A10Rik, PZR
- Chromosome Location
-
Chr.1: 165592181 - 165634558 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK210564.1 |
- |
1 - 2 |
54 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
adenylate cyclase 10 |
Adcy10 |
4930431D04Rik, 4931412F17, Sacy, sAC |
-31.7kb
|
Anti-sense |
Chr. - 165483869 - 165576778 on Build GRCm38 |
Downstream |
RCSD domain containing 1 |
Rcsd1 |
A430105K13Rik, BC025872 |
35.0kb
|
Sense |
Chr. - 165648945 - 165710063 on Build GRCm38 |
View Full Product Details