Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Arhgap32
- Entrez Gene ID
-
330914
- Gene Name
- Rho GTPase activating protein 32
- Gene Aliases
-
3426406O18Rik, Gc-gap, Grit, Px-rics, Rics, mKIAA0712, p200Rhogap, p250Gap
- Chromosome Location
-
Chr.9: 32016882 - 32268443 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014385.1 |
- |
1 - 1 |
1047 |
110 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
BarH-like homeobox 2 |
Barx2 |
2310006E12Rik, Barx2b |
-351.4kb
|
Anti-sense |
Chr. - 31846044 - 31913285 on Build GRCm38 |
Downstream |
potassium inwardly-rectifying channel, subfamily J, member 5 |
Kcnj5 |
GIRK4, Kir3.4 |
48.8kb
|
Anti-sense |
Chr. - 32314706 - 32344389 on Build GRCm38 |
View Full Product Details