Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Il15
- Entrez Gene ID
-
16168
- Gene Name
- interleukin 15
- Gene Aliases
-
AI503618, IL-15
- Chromosome Location
-
Chr.8: 82331624 - 82403227 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB022307.1 |
- |
1 - 1 |
71 |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
inositol polyphosphate-4-phosphatase, type II |
Inpp4b |
E130107I17Rik |
-191.3kb
|
Anti-sense |
Chr. - 81342342 - 82152019 on Build GRCm38 |
Downstream |
RIKEN cDNA 9530004M14 gene |
9530004M14Rik |
|
59.9kb
|
Anti-sense |
Chr. - 82403316 - 82403720 on Build GRCm38 |
View Full Product Details