Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK155243.1 - 2345 Chr. - - [-] on Build 118
GenBankmRNA BY531039.1 - 1 132 Chr. 14 - 27351512 - 27351967 [+] on Build GRCm38 118
GenBankmRNA BB464983.2 - 1 142 Chr. 14 - 27351499 - 27352168 [+] on Build GRCm38 118


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream interleukin 17 receptor D Il17rd 2810004A10Rik, AI428510, Sef, Sef-S -244.2kb Sense Chr. - 27038801 - 27107286 on Build GRCm38
Downstream family with sequence similarity 208, member A Fam208a 4732418E12, 4933409E02Rik, D14Abb1e 74.5kb Sense Chr. - 27426658 - 27483555 on Build GRCm38

View Full Product Details