The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160043.1 | NP_001153515.1 | 1 - 2 | 496 | 65 |
GenBank mRNA | CA548348.1 | - | 1 - 2 | 508 | 65 |