The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163621.1 | NP_001157093.1 | 3 - 4 | 289 | 100 |
NM_028010.1 | NP_082286.1 | 3 - 4 | 257 | 100 | |
GenBank mRNA | AK010208.1 | - | 3 - 4 | 258 | 100 |
AK144563.1 | - | 3 - 4 | 290 | 100 |