The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164797.1 | NP_001158269.1 | 5 - 6 | 1066 | 66 |
GenBank mRNA | AK132457.1 | - | 5 - 6 | 1067 | 66 |