Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il1rl1
Entrez Gene ID
17082
Gene Name
interleukin 1 receptor-like 1
Gene Aliases
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2
Chromosome Location
Chr.1: 40404864 - 40465418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001294171.1 NP_001281100.1 8 - 8 1298 71
NM_010743.3 NP_034873.2 8 - 8 1465 71
GenBank mRNA AK033417.1 - 8 - 8 1318 71
M24843.1 - 8 - 8 1306 71
Y07519.1 - 8 - 8 1309 71

View Full Product Details