The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079869.1 | NP_001073338.1 | 4 - 4 | 1730 | 91 |
NM_010458.2 | NP_034588.2 | 2 - 2 | 1653 | 91 | |
GenBank mRNA | S35628.1 | - | 2 - 2 | 1653 | 91 |
S35738.1 | - | 4 - 4 | 1731 | 91 | |
X66177.1 | - | 2 - 2 | 1653 | 91 |