Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Copz2
Entrez Gene ID
56358
Gene Name
coatomer protein complex, subunit zeta 2
Gene Aliases
1110012D12Rik, zeta2-COP
Chromosome Location
Chr.11: 96849876 - 96861203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019877.2 NP_063930.1 2 - 3 190 103
GenBank mRNA AB040137.1 - 2 - 3 193 103
AB047850.1 - 2 - 3 203 103
AF237687.1 - 2 - 3 193 103
AK003622.1 - 2 - 3 113 103
AK187566.1 - 1 - 2 72 103
BC025122.1 - 2 - 3 179 103

View Full Product Details