The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014395.2 | NP_001014417.1 | 9 - 10 | 1321 | 91 |
GenBank mRNA | AK135808.1 | - | 7 - 8 | 1038 | 91 |
AK162068.1 | - | 5 - 6 | 829 | 91 | |
BC084593.1 | - | 9 - 10 | 1325 | 91 |