The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024148.1 | NP_001019319.1 | 2 - 3 | 766 | 98 |
GenBank mRNA | BC048920.1 | - | 1 - 2 | 767 | 98 |
BC058786.1 | - | 2 - 3 | 766 | 98 |