Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mup2
Entrez Gene ID
17841
Gene Name
major urinary protein 2
Gene Aliases
AA589603, Mup-2, Mup4
Chromosome Location
Chr.4: 60135913 - 60139872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045550.2 NP_001039015.1 5 - 6 587 95
NM_001286096.1 NP_001273025.1 4 - 5 453 95
NM_008647.4 NP_032673.3 5 - 6 587 95
GenBank mRNA AI194744.1 - - 95
AI255759.1 - 2 - 3 95
AW112687.1 - 1 - 2 95
AW112688.1 - 1 - 2 95
BC012259.1 - - 576 95
BC059097.1 - - 562 95
M16356.1 - - 575 95

Gene Symbol
Gm2083
Entrez Gene ID
100048885
Gene Name
major urinary protein LOC100048885
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134644.1 NP_001128116.1 5 - 6 582 95

Gene Symbol
LOC100048884
Entrez Gene ID
100048884
Gene Name
novel member of the major urinary protein (Mup) gene family
Gene Aliases
Mup6
Chromosome Location
Chr.4: 61670177 - 61674098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199333.1 NP_001186262.1 5 - 6 589 95
GenBank mRNA BC055353.1 - - 564 95

Gene Symbol
Mup1
Entrez Gene ID
17840
Gene Name
major urinary protein 1
Gene Aliases
Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
Chromosome Location
Chr.4: 60498012 - 60501960 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC013649.1 - - 562 95
BC037152.1 - - 452 95
BC099597.1 - - 565 95
M27609.1 - - 430 95
X03524.1 - - 430 95
X04115.1 - - 386 95

Gene Symbol
Mup10
Entrez Gene ID
100039008
Gene Name
major urinary protein 10
Gene Aliases
2610016E04Rik, Mup8
Chromosome Location
Chr.4: 60578260 - 60582155 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122647.1 NP_001116119.1 5 - 6 584 95
GenBank mRNA AI118190.1 - 2 - 3 95
AK011413.1 - 3 - 4 636 95

Gene Symbol
Mup11
Entrez Gene ID
100039028
Gene Name
major urinary protein 11
Gene Aliases
Gm12549, OTTMUSG00000007431
Chromosome Location
Chr.4: 60658466 - 60662395 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164526.1 NP_001157998.1 - 615 95
GenBank mRNA AI118205.1 - 2 - 3 95
AK013972.1 - 5 - 6 592 95

Gene Symbol
Mup14
Entrez Gene ID
100039116
Gene Name
major urinary protein 14
Gene Aliases
Gm13514, Mup12, OTTMUSG00000012493
Chromosome Location
Chr.4: 61300037 - 61303998 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199999.1 NP_001186928.1 5 - 6 626 95

Gene Symbol
Mup16
Entrez Gene ID
100039177
Gene Name
major urinary protein 16
Gene Aliases
-
Chromosome Location
Chr.4: 61515592 - 61519503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199936.1 NP_001186865.1 5 - 6 583 95
GenBank mRNA AW112504.1 - 2 - 3 95

Gene Symbol
Mup17
Entrez Gene ID
100039206
Gene Name
major urinary protein 17
Gene Aliases
Gm12557, MUP 17, OTTMUSG00000007480
Chromosome Location
Chr.4: 61591929 - 61595853 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001200006.1 NP_001186935.1 5 - 6 589 95

Gene Symbol
Mup19
Entrez Gene ID
100189605
Gene Name
major urinary protein 19
Gene Aliases
100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
Chromosome Location
Chr.4: 61778324 - 61782236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135127.2 NP_001128599.1 5 - 6 583 95
GenBank mRNA AK013259.1 - 5 - 6 583 95

Gene Symbol
Mup9
Entrez Gene ID
100038948
Gene Name
major urinary protein 9
Gene Aliases
Gm14076, Mup2, Mup6, OTTMUSG00000015595
Chromosome Location
Chr.4: 60418046 - 60421961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281979.1 NP_001268908.1 5 - 6 583 95
GenBank mRNA BI219655.1 - 5 - 6 564 95

View Full Product Details