Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pard6a
Entrez Gene ID
56513
Gene Name
par-6 family cell polarity regulator alpha
Gene Aliases
0710008C04Rik, 2610010A15Rik, PAR-6A, PAR6alpha, Par-6, Par6, Par6c, TAX40, Tip-40
Chromosome Location
Chr.8: 105701148 - 105703494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047435.2 NP_001040900.1 1 - 2 157 115
NM_001286345.1 NP_001273274.1 1 - 2 157 115
NM_019695.3 NP_062669.2 1 - 2 157 115
GenBank mRNA AF070970.1 - 1 - 2 153 115
AK158848.1 - 1 - 2 155 115
BC049593.1 - 1 - 2 134 115
BC061114.1 - 1 - 2 157 115
BG806503.1 - 1 - 2 82 115
BY280124.1 - 1 - 2 155 115

View Full Product Details