Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r21
- Entrez Gene ID
-
546912
- Gene Name
- vomeronasal 2, receptor 21
- Gene Aliases
-
EG546912
- Chromosome Location
-
Chr.6: 123492508 - 123533406 on Build GRCm38
- Gene Symbol
- Vmn2r20
- Entrez Gene ID
-
667180
- Gene Name
- vomeronasal 2, receptor 20
- Gene Aliases
-
EG667180
- Chromosome Location
-
Chr.6: 123385262 - 123418061 on Build GRCm38
- Gene Symbol
- Vmn2r22
- Entrez Gene ID
-
546913
- Gene Name
- vomeronasal 2, receptor 22
- Gene Aliases
-
EG546913
- Chromosome Location
-
Chr.6: 123609758 - 123650635 on Build GRCm38
View Full Product Details