The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008199.2 | NP_032225.2 | 5 - 6 | 1033 | 89 |
GenBank mRNA | AY989822.1 | - | 5 - 6 | 974 | 89 |
AY989823.1 | - | 5 - 6 | 933 | 89 | |
AY989824.1 | - | 4 - 5 | 731 | 89 | |
AY989825.1 | - | 4 - 5 | 698 | 89 | |
AY989826.1 | - | 4 - 5 | 731 | 89 | |
AY989827.1 | - | 4 - 5 | 705 | 89 | |
AY989829.1 | - | 3 - 4 | 307 | 89 | |
AY989832.1 | - | 3 - 4 | 227 | 89 | |
AY989833.1 | - | 2 - 3 | 185 | 89 | |
AY989834.1 | - | 2 - 3 | 104 | 89 | |
AY989838.1 | - | 5 - 6 | 1007 | 89 | |
AY989839.1 | - | 5 - 6 | 974 | 89 | |
AY989840.1 | - | 4 - 5 | 731 | 89 | |
AY989841.1 | - | 4 - 5 | 698 | 89 | |
AY989842.1 | - | 5 - 6 | 1007 | 89 | |
AY989843.1 | - | 5 - 6 | 974 | 89 | |
AY989844.1 | - | 5 - 6 | 1007 | 89 | |
AY989845.1 | - | 5 - 6 | 974 | 89 | |
AY989846.1 | - | 4 - 5 | 731 | 89 | |
AY989847.1 | - | 4 - 5 | 698 | 89 | |
AY989848.1 | - | 4 - 5 | 698 | 89 | |
AY989856.1 | - | 4 - 5 | 731 | 89 | |
AY989857.1 | - | 4 - 5 | 698 | 89 |