Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmnn
Entrez Gene ID
57441
Gene Name
geminin
Gene Aliases
AW546347, Gem
Chromosome Location
Chr.13: 24751845 - 24761937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020567.2 NP_065592.1 4 - 5 395 82
GenBank mRNA AF068780.1 - 4 - 5 542 82
AK131959.1 - 4 - 5 369 82
BC020061.1 - 4 - 5 547 82

View Full Product Details