Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txndc2
Entrez Gene ID
213272
Gene Name
thioredoxin domain containing 2 (spermatozoa)
Gene Aliases
AU021712, Sptrx-1, Sptrx1, Trx4
Chromosome Location
Chr.17: 65637505 - 65643550 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146002.1 NP_001139474.1 2 - 3 375 102
NM_153519.2 NP_705739.2 1 - 2 203 102
GenBank mRNA AF196282.2 - 1 - 2 203 102
BC060981.1 - 2 - 3 340 102
BU938074.1 - 2 - 3 378 102

View Full Product Details