The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161366.1 | NP_001154838.1 | 1 - 2 | 6196 | 89 |
GenBank mRNA | BC145247.1 | - | 1 - 2 | 6162 | 89 |