Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrd6
Entrez Gene ID
210510
Gene Name
tudor domain containing 6
Gene Aliases
AB097085, Tdr2
Chromosome Location
Chr.17: 43615334 - 43631350 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161366.1 NP_001154838.1 4 - 5 6474 104
NM_198418.2 NP_940810.2 4 - 5 6471 104
GenBank mRNA AB097085.1 - 4 - 5 6471 104
BC138353.1 - 5 - 6 5859 104
BC145247.1 - 4 - 5 6440 104

View Full Product Details