The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008178.2 | NP_032204.1 | 1 - 2 | 604 | 78 |
GenBank mRNA | AB221618.1 | - | 1 - 2 | 414 | 78 |
BC137769.1 | - | 1 - 2 | 573 | 78 | |
U21224.1 | - | 1 - 2 | 604 | 78 |