Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd7
Entrez Gene ID
12516
Gene Name
CD7 antigen
Gene Aliases
-
Chromosome Location
Chr.11: 121036747 - 121039478 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009854.2 NP_033984.1 1 - 2 184 100
GenBank mRNA AK136444.1 - 1 - 2 138 100
AK154339.1 - 1 - 2 139 100
D10329.1 - 1 - 2 124 100

View Full Product Details