Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csn2
Entrez Gene ID
12991
Gene Name
casein beta
Gene Aliases
Csnb
Chromosome Location
Chr.5: 87692619 - 87699429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286020.1 NP_001272949.1 1 - 2 53 68
NM_001286022.1 NP_001272951.1 1 - 2 53 68
NM_001286023.1 NP_001272952.1 1 - 2 53 68
NM_001286024.1 NP_001272953.1 1 - 2 53 68
NM_009972.2 NP_034102.1 1 - 2 53 68
GenBank mRNA AK021324.1 - 1 - 2 50 68
AK021328.1 - 1 - 2 50 68
AK052803.1 - 1 - 2 50 68
AK052805.1 - 1 - 2 50 68
AK052822.1 - 1 - 2 50 68
AK085678.1 - 1 - 2 50 68
AK085694.1 - 1 - 2 50 68
AK085698.1 - 1 - 2 50 68
AK085729.1 - 1 - 2 50 68
AK085756.1 - 1 - 2 50 68
AK142616.1 - 1 - 2 50 68
AK142620.1 - 1 - 2 50 68
AK142631.1 - 1 - 2 50 68
AK142635.1 - 1 - 2 50 68
AK164747.1 - 1 - 2 50 68
AK164761.1 - 1 - 2 50 68
AK164791.1 - 1 - 2 50 68
AK164792.1 - 1 - 2 47 68
AK164793.1 - 1 - 2 47 68
AK164794.1 - 1 - 2 50 68
X04490.1 - 1 - 2 49 68

View Full Product Details