Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Prl2c2
- Entrez Gene ID
-
18811
- Gene Name
- prolactin family 2, subfamily c, member 2
- Gene Aliases
-
MRP-1, PLF-1, Plf, Plf1
- Chromosome Location
-
Chr.13: 12996120 - 13005330 on Build GRCm38
- Gene Symbol
- Prl2c3
- Entrez Gene ID
-
18812
- Gene Name
- prolactin family 2, subfamily c, member 3
- Gene Aliases
-
MRP-2, MRP-3, Mrpplf3, PLF-2, PLF-3, Plf2, Prl2c4, mrp/plf3
- Chromosome Location
-
Chr.13: 12790816 - 12800093 on Build GRCm38
- Gene Symbol
- Prl2c4
- Entrez Gene ID
-
26421
- Gene Name
- prolactin family 2, subfamily c, member 4
- Gene Aliases
-
MRP-3, Mrpplf3, PLF-3, mrp/plf3
- Chromosome Location
-
-
View Full Product Details