The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289465.1 | NP_001276394.1 | 2 - 3 | 1878 | 76 |
NM_001289466.1 | NP_001276395.1 | 2 - 3 | 1134 | 76 | |
NM_011441.5 | NP_035571.1 | 2 - 3 | 1878 | 76 | |
GenBank mRNA | AK004781.1 | - | 2 - 3 | 939 | 76 |
AK013848.1 | - | 1 - 2 | 163 | 76 | |
AK076707.1 | - | 2 - 3 | 1881 | 76 |