The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025453.3 | NP_079729.1 | 2 - 3 | 286 | 105 |
GenBank mRNA | AK007532.1 | - | 2 - 3 | 105 | |
AK017195.1 | - | 2 - 3 | 287 | 105 | |
AK017209.1 | - | 2 - 3 | 295 | 105 | |
BC117736.2 | - | 2 - 3 | 278 | 105 |