The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027988.4 | NP_082264.2 | 5 - 6 | 1414 | 93 |
GenBank mRNA | AF399754.1 | - | 4 - 5 | 400 | 93 |
AF539797.1 | - | 4 - 5 | 400 | 93 | |
AK009605.1 | - | 4 - 5 | 509 | 93 | |
AK088226.1 | - | 5 - 6 | 1373 | 93 | |
AK157993.1 | - | 5 - 6 | 1415 | 93 | |
BC019525.1 | - | 4 - 5 | 426 | 93 | |
BC119527.1 | - | 4 - 5 | 502 | 93 | |
BC127235.1 | - | 4 - 5 | 502 | 93 | |
BC127236.1 | - | 4 - 5 | 502 | 93 |