The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133893.3 | NP_598654.1 | 5 - 6 | 1119 | 87 |
GenBank mRNA | AB067532.1 | - | 5 - 6 | 1073 | 87 |
AK054465.1 | - | 5 - 6 | 1087 | 87 | |
AK135783.1 | - | 4 - 5 | 633 | 87 | |
AK136001.1 | - | 5 - 6 | 1087 | 87 | |
AY055829.1 | - | 5 - 6 | 1081 | 87 | |
BC052835.1 | - | 5 - 6 | 1083 | 87 |