Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Tuba3a
Entrez Gene ID
22144
Gene Name
tubulin, alpha 3A
Gene Aliases
AI528779, M[a]3, Tuba3
Chromosome Location
Chr.6: 125278274 - 125286042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009446.2 NP_033472.1 1 - 1 52 84
GenBank mRNA AK132775.1 - 1 - 1 52 84

View Full Product Details