Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Mrgpra5
Entrez Gene ID
404235
Gene Name
MAS-related GPR, member A5
Gene Aliases
MrgA5
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207534.1 NP_997417.1 - 650 97

View Full Product Details