The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286033.1 | NP_001272962.1 | 2 - 3 | 263 | 72 |
NM_001286034.1 | NP_001272963.1 | 2 - 3 | 258 | 72 | |
NM_007941.3 | NP_031967.2 | 2 - 3 | 263 | 72 | |
GenBank mRNA | AK146462.1 | - | 2 - 3 | 192 | 72 |
AK165552.1 | - | 2 - 3 | 191 | 72 | |
AK167577.1 | - | 2 - 3 | 226 | 72 | |
BC046279.1 | - | 2 - 3 | 187 | 72 | |
CJ166382.1 | - | 2 - 3 | 256 | 72 | |
D10475.1 | - | 2 - 3 | 266 | 72 |