Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- C4bp-ps1
- Entrez Gene ID
-
12270
- Gene Name
- complement component 4 binding protein, pseudogene 1
- Gene Aliases
-
C4bpb
- Chromosome Location
-
Chr.1: 130670202 - 130681571 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028304.1 |
|
1 - 2 |
235 |
77 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
complement component 4 binding protein |
C4bp |
AI195242, C4bpa |
-16.2kb
|
Sense |
Chr. - 130635921 - 130661618 on Build GRCm38 |
Downstream |
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 |
Pfkfb2 |
4930568D07Rik |
7.5kb
|
Sense |
Chr. - 130689043 - 130729253 on Build GRCm38 |
View Full Product Details