The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020611.4 | NP_065636.2 | 1 - 2 | 278 | 63 |
GenBank mRNA | AI553072.1 | - | 1 - 2 | 280 | 63 |
AK040198.1 | - | 1 - 2 | 246 | 63 | |
AK075635.1 | - | 1 - 2 | 274 | 63 | |
AK138681.1 | - | 1 - 2 | 270 | 63 | |
AK139290.1 | - | 1 - 2 | 275 | 63 | |
AK162527.1 | - | 1 - 2 | 246 | 63 | |
BC014771.1 | - | 2 - 2 | 278 | 63 | |
BC039564.1 | - | 1 - 2 | 231 | 63 | |
BC145647.1 | - | 1 - 2 | 274 | 63 |