The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_045685.1 | 2 - 3 | 253 | 104 | |
GenBank mRNA | AK006470.1 | - | 2 - 3 | 254 | 104 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | secretoglobin, family 1B, member 20 | Scgb1b20 | EG545948, Gm5895 | -250.6kb | Anti-sense | Chr. - 33373234 - 33374559 on Build GRCm38 |
Downstream | secretoglobin, family 2B, member 24 | Scgb2b24 | Abpbg24, Abpz, C2b, Scgb2b3 | 110.6kb | Sense | Chr. - 33737193 - 33739295 on Build GRCm38 |