Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Tmem181c-ps
- Entrez Gene ID
-
100040525
- Gene Name
- transmembrane protein 181C, pseudogene
- Gene Aliases
-
100040596, ENSMUSG00000079729, ENSMUSG00000079731, Tmem181d-ps
- Chromosome Location
-
Chr.17: 6610103 - 6620925 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_028305.2 |
|
2 - 3 |
661 |
100 |
GenBank mRNA |
AK045744.1 |
- |
2 - 3 |
662 |
100 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG276789.1 |
- |
1 - 2 |
89 |
|
Chr. 17 - 6613777 - 6620187 [-] on Build GRCm38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dynein light chain Tctex-type 1C |
Dynlt1c |
100040603, Dynlt1d |
-11.2kb
|
Anti-sense |
Chr. - 6601779 - 6607891 on Build GRCm38 |
Downstream |
dynein light chain Tctex-type 1F |
Dynlt1f |
100040631, Dynlt1e |
26.2kb
|
Sense |
Chr. - 6646606 - 6655939 on Build GRCm38 |
View Full Product Details