Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rchy1
- Entrez Gene ID
-
68098
- Gene Name
- ring finger and CHY zinc finger domain containing 1
- Gene Aliases
-
ARNIP, CHIMP, Pirh2, Zfp363, Znf363
- Chromosome Location
-
Chr.5: 91948842 - 91963224 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073442.1 |
|
2 - 3 |
430 |
128 |
GenBank mRNA |
AK015870.1 |
- |
2 - 3 |
431 |
128 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY715421.1 |
- |
2 - 3 |
431 |
|
Chr. 5 - 91951855 - 91962556 [-] on Build GRCm38 |
128 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
RIKEN cDNA 4930570G05 gene |
4930570G05Rik |
|
-44.1kb
|
Sense |
Chr. - 91911398 - 91913806 on Build GRCm38 |
Downstream |
THAP domain containing 6 |
Thap6 |
4930534K21Rik, AI448650 |
1.8kb
|
Anti-sense |
Chr. - 91962882 - 91972137 on Build GRCm38 |
View Full Product Details