Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Chchd7
- Entrez Gene ID
-
66433
- Gene Name
- coiled-coil-helix-coiled-coil-helix domain containing 7
- Gene Aliases
-
1810049H20Rik
- Chromosome Location
-
Chr.4: 3938860 - 3951382 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104355.1 |
|
8 - 9 |
1131 |
96 |
GenBank mRNA |
AK145105.1 |
- |
8 - 9 |
1106 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
pleiomorphic adenoma gene 1 |
Plag1 |
|
-10.6kb
|
Anti-sense |
Chr. - 3898769 - 3938434 on Build GRCm38 |
Downstream |
short chain dehydrogenase/reductase family 16C, member 5 |
Sdr16c5 |
Rdhe2, Scdr9 |
44.6kb
|
Anti-sense |
Chr. - 3995942 - 4019663 on Build GRCm38 |
View Full Product Details