Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lmtk3
Entrez Gene ID
381983
Gene Name
lemur tyrosine kinase 3
Gene Aliases
BC059845, aatyk3
Chromosome Location
Chr.7: 45783743 - 45806891 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110987.1 9 - 10 942 68
GenBank mRNA BC059845.1 - 9 - 10 1151 68


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream sulfotransferase family, cytosolic, 2B, member 1 Sult2b1 AI326997, BB173635, ST2B1, SULT2B -32.8kb Anti-sense Chr. - 45721174 - 45759555 on Build GRCm38
Downstream cytohesin 2 Cyth2 ARNO, CLM2, Pscd2 14.0kb Anti-sense Chr. - 45806637 - 45814316 on Build GRCm38

View Full Product Details