Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPARC
Entrez Gene ID
100307048
Gene Name
secreted protein acidic and cysteine rich
Gene Aliases
-
Chromosome Location
Chr.5: 65593486 - 65615912 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166202.1 NP_001159674.1 1 - 2 52 92
XM_012177565.3 XP_012032955.1 2 - 3 232 92
GenBank mRNA GQ221054.1 - 1 - 2 52 92

View Full Product Details