Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA3
Entrez Gene ID
101103257
Gene Name
NADH:ubiquinone oxidoreductase subunit A3
Gene Aliases
-
Chromosome Location
Chr.14: 68196395 - 68199486 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015437.4 XP_004015486.1 3 - 4 232 61

View Full Product Details