Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM4
Entrez Gene ID
101117121
Gene Name
RNA binding motif protein 4
Gene Aliases
-
Chromosome Location
Chr.21: 46438514 - 46448327 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019707.4 XP_004019756.2 3 - 4 3506 57

View Full Product Details