Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR13
Entrez Gene ID
101103886
Gene Name
proline rich 13
Gene Aliases
-
Chromosome Location
Chr.3: 142593968 - 142597430 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006298.4 XP_004006347.1 3 - 4 513 100

Gene Symbol
LOC114117553
Entrez Gene ID
114117553
Gene Name
proline-rich protein 13-like
Gene Aliases
-
Chromosome Location
Chr.13: 17124687 - 17125682 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976541.1 XP_027832342.1 1 - 1 491 100

View Full Product Details