Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATF3
Entrez Gene ID
101116073
Gene Name
activating transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.17: 38274586 - 38287479 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013898.3 XP_004013947.2 1 - 2 3056 79
XM_015099372.2 XP_014954858.1 2 - 3 433 79

View Full Product Details