Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFP1
Entrez Gene ID
101111663
Gene Name
mitochondrial fission process 1
Gene Aliases
-
Chromosome Location
Chr.17: 78705702 - 78713702 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012098057.3 XP_011953447.2 2 - 3 693 109
XM_012098058.3 XP_011953448.2 2 - 3 698 109

View Full Product Details