Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BANF1
Entrez Gene ID
101112433
Gene Name
BAF nuclear assembly factor 1
Gene Aliases
-
Chromosome Location
Chr.21: 45879754 - 45881788 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959814.1 XP_027815615.1 2 - 3 342 98

View Full Product Details