Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNA3
Entrez Gene ID
101115414
Gene Name
ephrin A3
Gene Aliases
-
Chromosome Location
Chr.1: 111446821 - 111455586 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002571.4 XP_004002620.1 2 - 3 531 71

View Full Product Details